Dimethyl-3-nitrophthalate ≥98%
Fornitore: Thermo Fisher Scientific
Synonyms:
Dimethyl 3-nitrophthalate
Purity: 98%
CAS No.: 13365-26-9
Molecular Formula: C10H9NO6
Formula Weight: 239.19
Melting Point: 67-70°
Beilstein Reference: 1885570
Formula:
C₁₀H₉NO₆ Peso molecolare: 239.19 g/mol Punto di fusione: 67…70 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00017184 CAS Number: 13365-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...