Stampa…

2'-Ethoxyacetanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: N-Acetyl-o-phenetidine

A0103-25G A0103-500G
TCIAA0103-25GEA 68 EUR
TCIAA0103-25G TCIAA0103-500G
2'-Ethoxyacetanilide ≥98.0% (by GC)
2'-Ethoxyacetanilide

Other Product Names / Synonyms: N-Acetyl-2-ethoxyaniline, 2'-Ethoxyacetanilide

Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,22 g/mol
MDL Number: MFCD00059228
CAS Number: 581-08-8
EINECS: 209-460-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=52fd71a739378a0ceaf3a0884a573872b0eb0d31&vid=52fd71a739378a0ceaf3a0884a573872b0eb0d31&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 81.0 deg-C
Solubility in Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR