Stampa…

2'-Methyl-3'-nitroacetanilide ≥98.0% (by GC)

Fornitore: TCI EUROPE

Synonyms: N-(2-Methyl-3-nitrophenyl)acetamide

M1651-25G M1651-5G
TCIAM1651-25GEA 140 EUR
TCIAM1651-25G TCIAM1651-5G
2'-Methyl-3'-nitroacetanilide ≥98.0% (by GC)
2'-Methyl-3'-nitroacetanilide
Formula: C₉H₁₀N₂O₃
Peso molecolare: 194,190 g/mol
MDL Number: MFCD01047363
CAS Number: 56207-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ecb851ff262eb0d19924297ecc5e8aa6b26450c&vid=2ecb851ff262eb0d19924297ecc5e8aa6b26450c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 161.0 to 165.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR