Phenyl-4-hydroxybenzoate ≥99.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Formula:
C₁₃H₁₀O₃ |
MDL Number:
MFCD00016467 CAS Number: 17696-62-7 EINECS: 241-698-9 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 180.0 to 183.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...