Stampa…

3-Amino-3-(4-methylphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)

Fornitore: TCI EUROPE

Synonyms: 3-Amino-3-p-tolyl-propionic acid, DL-3-Amino-3-p-tolyl-propionic acid

A2345-25G A2345-5G
TCIAA2345-25GEA 203 EUR
TCIAA2345-25G TCIAA2345-5G
3-Amino-3-(4-methylphenyl)propionic acid ≥98.0% (by HPLC, titration analysis)
3-Amino-3-(4-methylphenyl)propionic acid
Formula: C₁₀H₁₃NO₂
Peso molecolare: 179,2157 g/mol
Punto di fusione: 222°C
MDL Number: MFCD01863249
CAS Number: 68208-18-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=454faadc514e2ba3fda4aa92c944c7c3ab1c7473&vid=454faadc514e2ba3fda4aa92c944c7c3ab1c7473&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR