Pentachlorothioanisole ≥98.0% (by GC)
Fornitore: TCI
P1067-1G
P1067-5G
TCIAP1067-1GEA
48
EUR
TCIAP1067-1G
TCIAP1067-5G
Pentachlorothioanisole ≥98.0% (by GC)
Pentachlorothioanisole
Formula:
C₇H₃Cl₅S |
MDL Number:
MFCD00013622 CAS Number: 1825-19-0 EINECS: 217-363-8 |
|
Risultati del test di specificazione
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 93.0 to 96.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...