Chlorobis(3,5-dimethylphenyl)phosphine tech. 90%, grado tecnico
Fornitore: Thermo Scientific
|
Pericolo
|
Synonyms:
Bis(3,5-dimethylphenyl)chlorophosphine
H25885.06
H25885.03
H25885.06EA
354
EUR
H25885.06
H25885.03
Chlorobis(3,5-dimethylphenyl)phosphine tech. 90%, grado tecnico
Chlorobis(3,5-dimethylphenyl)phosphine
Purity: 90%
CAS No.: 74289-57-9
Molecular Formula: C16H18ClP
Formula Weight: 276.75
UN No.: 3261
MDL No.: MFCD01630841
Formula:
C₁₆H₁₈ClP Peso molecolare: 276,75 g/mol Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD01630841 CAS Number: 74289-57-9 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...