3-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Hydroxy-3-methylbenzaldehyde
M2270-25G
M2270-5G
TCIAM2270-25GEA
230
EUR
TCIAM2270-25G
TCIAM2270-5G
3-Methylsalicylaldehyde ≥98.0% (by GC, titration analysis)
3-Methylsalicylaldehyde
Formula:
C₈H₈O₂ Peso molecolare: 136,1479 g/mol Punto di ebollizione: 209°C Punto di fusione: 17°C Flash Pt: 87°C |
MDL Number:
MFCD00040798 CAS Number: 824-42-0 |
Risultati del test di specificazione
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...