Stampa…

Sodium abietate ≥95.0% (by titrimetric analysis)

Fornitore: TCI

A0651-5G A0651-25G
TCIAA0651-5GEA 59 EUR
TCIAA0651-5G TCIAA0651-25G
Sodium abietate ≥95.0% (by titrimetric analysis)
Sodium abietate
Formula: C₂₀H₂₉NaO₂
MDL Number: MFCD00058941
CAS Number: 14351-66-7
EINECS: 238-313-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cecded9f5c9e19c7b6815b714e07b6a80ee95222&vid=cecded9f5c9e19c7b6815b714e07b6a80ee95222&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR