4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Doxazosin Related Compound C, 2-Chloro-4-amino-6,7-dimethoxyquinazoline
A2151-25G
A2151-5G
TCIAA2151-25GEA
143
EUR
TCIAA2151-25G
TCIAA2151-5G
4-Amino-2-chloro-6,7-dimethoxyquinazoline ≥98.0% (by HPLC, total nitrogen)
4-Amino-2-chloro-6,7-dimethoxyquinazoline
Formula:
C₁₀H₁₀ClN₃O₂ Peso molecolare: 239,66 g/mol Punto di fusione: 302°C |
MDL Number:
MFCD00051734 CAS Number: 23680-84-4 EINECS: 245-821-7 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...