N-Fmoc-glycine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI
Formula:
C₁₇H₁₅NO₄ |
MDL Number:
MFCD00037140 CAS Number: 29022-11-5 EINECS: 249-373-3 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 175.0 to 178.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...