4-Nitro-2,3-lutidine-N-oxide ≥98.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide
D2905-25G
D2905-5G
TCIAD2905-25GEA
42
EUR
TCIAD2905-25G
TCIAD2905-5G
4-Nitro-2,3-lutidine-N-oxide ≥98.0% (by titrimetric analysis)
4-Nitro-2,3-lutidine-N-oxide
Formula:
C₇H₈N₂O₂ Peso molecolare: 168,15 g/mol |
MDL Number:
MFCD00065172 CAS Number: 37699-43-7 |
Risultati del test di specificazione
Appearance | White to Light yellow to Green powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Solubility in hot Water | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...