Stampa…

2-Chloro-6-methylisonicotinic acid ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 2-Chloro-6-methylpyridine-4-carboxylic acid

C2486-1G C2486-5G
TCIAC2486-1GEA 32 EUR
TCIAC2486-1G TCIAC2486-5G
2-Chloro-6-methylisonicotinic acid ≥98.0% (by GC)
2-Chloro-6-methylisonicotinic acid
Formula: C₇H₆ClNO₂
Peso molecolare: 171,58 g/mol
MDL Number: MFCD00052830
CAS Number: 25462-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef9335fc66faaec74e91b1c6f8a5e48bb4bcc927&vid=ef9335fc66faaec74e91b1c6f8a5e48bb4bcc927&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR