2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
2-Mercaptopyridinen-oxide sodiumsalt, 2-Mercaptopyridine N-oxide sodium
M0632-25G
M0632-500G
M0632-100G
TCIAM0632-25GEA
21
EUR
TCIAM0632-25G
TCIAM0632-500G
TCIAM0632-100G
2-Mercaptopyridine-N-oxide sodium salt 40% ∼3.3 mol/L in water
2-Mercaptopyridine-N-oxide sodium salt
Formula:
C₅H₄NNaOS Peso molecolare: 149,15 g/mol |
MDL Number:
MFCD01941547 CAS Number: 3811-73-2 EINECS: 223-296-5 Merck Index: 14,07994 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange clear liquid |
Concentration (Iodometric titration) | 38.0 to 42.0 w/w% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...