Stampa…

4-Cyanopyridine-N-oxide ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-Cyanopyridine 1-oxide

C0765-25G C0765-500G
TCIAC0765-25GEA 71 EUR
TCIAC0765-25G TCIAC0765-500G
4-Cyanopyridine-N-oxide ≥98.0% (by titrimetric analysis)
4-Cyanopyridine-N-oxide
Formula: C₆H₄N₂O
Peso molecolare: 120,1088 g/mol
MDL Number: MFCD00006205
CAS Number: 14906-59-3
EINECS: 238-974-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5242078d1808a6d57bf85915fd526568b30d6fd9&vid=5242078d1808a6d57bf85915fd526568b30d6fd9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 227.0 to 232.0 deg-C
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR