2-(Bromoacetyl)naphthalene ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
Synonyms:
α-Bromo-2'-acetonaphthone, 2-Bromo-2'-acetonaphthone
B1259-25G
B1259-5G
TCIAB1259-25GEA
71
EUR
TCIAB1259-25G
TCIAB1259-5G
2-(Bromoacetyl)naphthalene ≥98.0% (by GC)
2-(Bromoacetyl)naphthalene
Formula:
C₁₂H₉BrO Peso molecolare: 249,11 g/mol |
MDL Number:
MFCD00004109 CAS Number: 613-54-7 EINECS: 210-348-7 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 85.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...