Trimanganese diphosphide ≥99% (metals basis)
Fornitore: Thermo Scientific
Synonyms:
Manganese phosphide
14020.04
14020.09
14020.04EA
33.9
EUR
14020.04
14020.09
Trimanganese diphosphide ≥99% (metals basis)
Trimanganese diphosphide
Purity: 99%
CAS No.: 12263-33-1
Molecular Formula: Mn3P2 (mixture of MnP & Mn2P)
Formula Weight: 226.75
Physical Form: -100 Mesh Powder
Odour: Odourless
MDL No.: MFCD00064736
Formula:
Mn₃P₂ Peso molecolare: 226,75 g/mol Punto di fusione: 1095 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00064736 CAS Number: 12263-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...