Stampa…

Tris(trimethylsilyl)borate ≥97.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Tris(trimethylsiloxy)borate

T2543-25G T2543-100G
TCIAT2543-25GEA 69 EUR
TCIAT2543-25G TCIAT2543-100G
Tris(trimethylsilyl)borate ≥97.0% (by titrimetric analysis)
Tris(trimethylsilyl)borate
Formula: C₉H₂₇BO₃Si₃
Peso molecolare: 278,376 g/mol
Punto di ebollizione: 180°C
Punto di fusione: –35°C
Flash Pt: 41°C
MDL Number: MFCD00051588
CAS Number: 4325-85-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a221430e4488008624fc9fd6688da5eedc8da58a&vid=a221430e4488008624fc9fd6688da5eedc8da58a&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR