Salicylaldehyde azine ≥97%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Synonyms:
Salicylaldazine
Purity: 97%
CAS No.: 959-36-4
Molecular Formula: C14H12N2O2
Formula Weight: 240.26
Melting Point: 216-222°
Beilstein Reference: 1913597
Formula:
C₁₄H₁₂N₂O₂ Punto di fusione: 216…222 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00043496 CAS Number: 959-36-4 EINECS: 213-500-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...