Cyclopropyl phenyl sulphide ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Cyclopropyl(phenyl)sulphane
C1014-25G
C1014-5G
TCIAC1014-25GEA
242
EUR
TCIAC1014-25G
TCIAC1014-5G
Cyclopropyl phenyl sulphide ≥98.0% (by GC)
Cyclopropyl phenyl sulphide
Formula:
C₉H₁₀S Peso molecolare: 150,24 g/mol Punto di ebollizione: 90°C Flash Pt: 91°C |
MDL Number:
MFCD00009684 CAS Number: 14633-54-6 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...