Methyl-4-methoxysalicylate ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-hydroxy-4-methoxybenzoate
M1797-25G
M1797-5G
TCIAM1797-25GEA
82
EUR
TCIAM1797-25G
TCIAM1797-5G
Methyl-4-methoxysalicylate ≥99.0% (by GC)
Methyl-4-methoxysalicylate
Formula:
C₉H₁₀O₄ Peso molecolare: 182,18 g/mol Punto di ebollizione: 134°C |
MDL Number:
MFCD00008424 CAS Number: 5446-02-6 EINECS: 226-657-5 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 48.0 to 52.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...