Stampa…

Methyl-3-methoxysalicylate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Methyl 2-hydroxy-3-methoxybenzoate

M2167-25G M2167-5G
TCIAM2167-25GEA 239 EUR
TCIAM2167-25G TCIAM2167-5G
Methyl-3-methoxysalicylate ≥98.0% (by GC)
Methyl-3-methoxysalicylate
Formula: C₉H₁₀O₄
Peso molecolare: 182,1700 g/mol
MDL Number: MFCD00239511
CAS Number: 6342-70-7
EINECS: 228-737-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aab69d91af148335f4d7eae803d3689c59c7b5aa&vid=aab69d91af148335f4d7eae803d3689c59c7b5aa&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 66.0 to 70.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR