Stampa…

2-Chloro-5-iodobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

C2212-1G C2212-5G
TCIAC2212-1GEA 28 EUR
TCIAC2212-1G TCIAC2212-5G
2-Chloro-5-iodobenzonitrile ≥98.0% (by GC)
2-Chloro-5-iodobenzonitrile
Formula: C₇H₃ClIN
Peso molecolare: 263,4629 g/mol
MDL Number: MFCD00672973
CAS Number: 289039-29-8
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7964f41bc6c3a8ef9d0fde49510084e7e0c698a6&vid=7964f41bc6c3a8ef9d0fde49510084e7e0c698a6&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR