Stampa…

5-Bromo-2-iodobenzonitrile ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

B2215-1G B2215-5G
TCIAB2215-1GEA 24 EUR
TCIAB2215-1G TCIAB2215-5G
5-Bromo-2-iodobenzonitrile ≥98.0% (by GC)
5-Bromo-2-iodobenzonitrile
Formula: C₇H₃BrIN
Peso molecolare: 307,92 g/mol
MDL Number: MFCD00672950
CAS Number: 121554-10-7
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6169aeb43ed6607134f39a9c1d438bf2af507cc&vid=e6169aeb43ed6607134f39a9c1d438bf2af507cc&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 120.0 to 124.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR