Stampa…

3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98.0% (by GC) stabilizzato

Fornitore: TCI EUROPE
Attenzione

Synonyms: 3-(1,1,1,5,5,5-Hexamethyl-3-((trimethylsilyl)oxy)trisiloxan-3-yl)propyl methacrylate, 3-(Methacryloyloxy)propyltris(trimethylsiloxy)silane

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

M1324-100ML M1324-25ML
TCIAM1324-100MLEA 144 EUR
TCIAM1324-100ML TCIAM1324-25ML
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate ≥98.0% (by GC) stabilizzato
3-[3,3,3-Trimethyl-1,1-bis[(trimethylsilyl)oxy]disiloxanyl]propyl methacrylate
Formula: C₁₆H₃₈O₅Si₄
Peso molecolare: 422,82 g/mol
Flash Pt: 200°C
MDL Number: MFCD00053871
CAS Number: 17096-07-0
EINECS: 241-165-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66d554aa93be22da87365d95a5221108f2bf9c09&vid=66d554aa93be22da87365d95a5221108f2bf9c09&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR