Stampa…

Potassium vinyltrifluoroborate ≥97.0% (W)

Fornitore: TCI
Attenzione

Synonyms: Potassium trifluoro(vinyl)borate

P1479-1G P1479-5G P1479-25G
TCIAP1479-1GEA 16 EUR
TCIAP1479-1G TCIAP1479-5G TCIAP1479-25G
Potassium vinyltrifluoroborate ≥97.0% (W)
Potassium vinyltrifluoroborate
Formula: C₂H₃BF₃K
MDL Number: MFCD02093335
CAS Number: 13682-77-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73787380c4bc7729d562386a28a432121a5476b2&vid=73787380c4bc7729d562386a28a432121a5476b2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (Gravimetric) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR