N,N'-Bis(salicylidene)ethylenediamine ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Ethylenebis(salicylimine), 2,2'-((Ethane-1,2-diylbis(azanylylidene))bis(methanylylidene))diphenol, SALEN
H0199-25G
H0199-500G
TCIAH0199-25GEA
24
EUR
TCIAH0199-25G
TCIAH0199-500G
N,N'-Bis(salicylidene)ethylenediamine ≥99.0% (by titrimetric analysis)
N,N'-Bis(salicylidene)ethylenediamine
Formula:
C₁₆H₁₆N₂O₂ Peso molecolare: 268,31 g/mol |
MDL Number:
MFCD00002244 CAS Number: 94-93-9 EINECS: 202-376-3 |
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 126.0 to 128.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...