Stampa…

3-(3,4-Dimethoxybenzoyl)propionic acid ≥98%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 4-(3,4-Dimethoxyphenyl)-4-oxobutanoic acid

H25806.14 H25806.06
H25806.14EA 322 EUR
H25806.14 H25806.06
3-(3,4-Dimethoxybenzoyl)propionic acid ≥98%
3-(3,4-Dimethoxybenzoyl)propionic acid

Purity: 98% CAS No.: 5333-34-6 Molecular Formula: C12H14O5 Formula Weight: 238.24 Melting Point: 159-161° MDL No.: MFCD00033286

Formula: C₁₂H₁₄O₅
Peso molecolare: 238,24 g/mol
Punto di fusione: 159…161 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00033286
CAS Number: 5333-34-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR