3-Bromo-9-ethylcarbazole ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3-Bromo-9-ethyl-9H-carbazole
B3459-1G
B3459-5G
TCIAB3459-1GEA
67
EUR
TCIAB3459-1G
TCIAB3459-5G
3-Bromo-9-ethylcarbazole ≥98.0% (by GC)
3-Bromo-9-ethylcarbazole
Formula:
C₁₄H₁₂BrN Peso molecolare: 274,1558 g/mol |
MDL Number:
MFCD00092392 CAS Number: 57102-97-3 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 82.0 to 86.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...