Diethylcyanamide 99+%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
200220100
200220500
ACRO200220100EA
102
EUR
ACRO200220100
ACRO200220500
Diethylcyanamide 99+%
Diethylcyanamide
Formula:
C₅H₁₀N₂ Flash Pt: 69 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00001769 CAS Number: 617-83-4 EINECS: 210-532-7 UN: 3276 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4210 to 1.4240 (20°C, 589 nm) |
GC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...