Ethyl isocyanoacetate ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Pericolo
|
I0562-25G
I0562-5G
TCIAI0562-25GEA
194
EUR
TCIAI0562-25G
TCIAI0562-5G
Ethyl isocyanoacetate ≥97.0% (by GC)
Ethyl isocyanoacetate
Formula:
C₅H₇NO₂ Peso molecolare: 113,12 g/mol Punto di ebollizione: 90°C Flash Pt: 84°C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00000007 CAS Number: 2999-46-4 EINECS: 221-077-9 UN: 2810 ADR: 6.1,III |
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...