Stampa…

Ethyl isocyanoacetate ≥98%

Fornitore: Thermo Scientific
Attenzione

L15379.03 L15379.06 L15379.14
L15379.03EA 54.2 EUR
L15379.03 L15379.06 L15379.14
Ethyl isocyanoacetate ≥98%
Ethyl isocyanoacetate

Purity: 98% CAS No.: 2999-46-4 Molecular Formula: CNCH2CO2CH2CH3 Formula Weight: 113.12 Storage Temperature: 2°C to 8°C Flash Point: 84°(183°F) Refractive Index: 1.4220 Boiling Point: 194-196° Beilstein Reference: 3588613 UN No.: 2810 MDL No.: MFCD00000007

Formula: C₅H₇NO₂
Peso molecolare: 113,12 g/mol
Punto di ebollizione: 194…196 °C
Densità: 1,035
Flash Pt: 84 °C (183 °F)
MDL Number: MFCD00000007
CAS Number: 2999-46-4
EINECS: 221-077-9
UN: 2810
ADR: 6.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR