Stampa…

Trimethylsilyl bromoacetate ≥97.0% (by GC)

Fornitore: TCI
Pericolo

B1578-25G B1578-5G
TCIAB1578-25GEA 124 EUR
TCIAB1578-25G TCIAB1578-5G
Trimethylsilyl bromoacetate ≥97.0% (by GC)
Trimethylsilyl bromoacetate
Formula: C₅H₁₁BrO₂Si
Peso molecolare: 211.13 g/mol
Punto di ebollizione: 83°C
Flash Pt: 28°C
MDL Number: MFCD00000187
CAS Number: 18291-80-0
EINECS: 242-165-3
UN: 2920
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=98d6ccdfeb0210115812e74e1a5b6f08aa33f8c2&vid=98d6ccdfeb0210115812e74e1a5b6f08aa33f8c2&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR