Stampa…

Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: Ethyl 2-amino-4-methylthiazole-5-carboxylate

E0898-5G E0898-1G
TCIAE0898-5GEA 74 EUR
TCIAE0898-5G TCIAE0898-1G
Ethyl-2-amino-4-methylthiazole-5-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl-2-amino-4-methylthiazole-5-carboxylate
Formula: C₇H₁₀N₂O₂S
Peso molecolare: 186,22 g/mol
MDL Number: MFCD00123414
CAS Number: 7210-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23644ab9b62fbbc459e2fb8ecf78aaafef8d396c&vid=23644ab9b62fbbc459e2fb8ecf78aaafef8d396c&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 176.0 to 180.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR