Stampa…

(Methoxymethyl)diphenylphosphine oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

M1210-25G M1210-5G
TCIAM1210-25GEA 279 EUR
TCIAM1210-25G TCIAM1210-5G
(Methoxymethyl)diphenylphosphine oxide ≥98.0% (by GC)
(Methoxymethyl)diphenylphosphine oxide
Formula: C₁₄H₁₅O₂P
Peso molecolare: 246,24 g/mol
MDL Number: MFCD00015451
CAS Number: 4455-77-0
EINECS: 224-704-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aecd345bd1b03c7e0195c3b8232974c44c35a837&vid=aecd345bd1b03c7e0195c3b8232974c44c35a837&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 113.0 to 117.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR