Stampa…

5-Iodo-2,4-dimethoxypyrimidine ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

I0531-1G I0531-5G
TCIAI0531-1GEA 85 EUR
TCIAI0531-1G TCIAI0531-5G
5-Iodo-2,4-dimethoxypyrimidine ≥98.0% (by GC)
5-Iodo-2,4-dimethoxypyrimidine
Formula: C₆H₇IN₂O₂
Peso molecolare: 266,04 g/mol
MDL Number: MFCD00090865
CAS Number: 52522-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c433f79fd119a450b31fa41fcc0da60a261d93c9&vid=c433f79fd119a450b31fa41fcc0da60a261d93c9&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 71.0 to 75.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR