Methyl-3-aminopyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-aminopyrazine-3-carboxylate
A1186-1G
A1186-25G
A1186-5G
TCIAA1186-1GEA
18
EUR
TCIAA1186-1G
TCIAA1186-25G
TCIAA1186-5G
Methyl-3-aminopyrazine-2-carboxylate ≥98.0% (by GC, titration analysis)
Methyl-3-aminopyrazine-2-carboxylate
Formula:
C₆H₇N₃O₂ Peso molecolare: 153,14 g/mol |
MDL Number:
MFCD00010097 CAS Number: 16298-03-6 EINECS: 240-387-5 |
Risultati del test di specificazione
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 170.0 to 173.0 deg-C |
Solubility in hot Methanol | very faint turbidity |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...