Methyl-2-pyrazinecarboxylate ≥98.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Methyl 2-pyrazinecarboxylate, Methyl pyrazine-2-carboxylate
M2015-25G
M2015-5G
TCIAM2015-25GEA
200
EUR
TCIAM2015-25G
TCIAM2015-5G
Methyl-2-pyrazinecarboxylate ≥98.0% (by GC)
Methyl-2-pyrazinecarboxylate
Formula:
C₆H₆N₂O₂ Peso molecolare: 138,13 g/mol Punto di ebollizione: 104°C |
MDL Number:
MFCD00014611 CAS Number: 6164-79-0 EINECS: 228-198-6 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...