Stampa…

Diphenylphosphine oxide ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

D2826-25G D2826-5G
TCIAD2826-25GEA 136 EUR
TCIAD2826-25G TCIAD2826-5G
Diphenylphosphine oxide ≥98.0% (by GC)
Diphenylphosphine oxide
Formula: C₁₂H₁₁OP
Peso molecolare: 202,20 g/mol
Punto di ebollizione: 105°C
MDL Number: MFCD00002079
CAS Number: 4559-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3187317474ec552b6f7ea058e681b21185f35917&vid=3187317474ec552b6f7ea058e681b21185f35917&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR