Phenylphosphinic acid ≥99.0% (by titrimetric analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
B0025-25G
B0025-500G
TCIAB0025-25GEA
16
EUR
TCIAB0025-25G
TCIAB0025-500G
Phenylphosphinic acid ≥99.0% (by titrimetric analysis)
Phenylphosphinic acid
Formula:
C₆H₇O₂P Peso molecolare: 142,092 g/mol |
MDL Number:
MFCD00002131 CAS Number: 1779-48-2 EINECS: 217-217-3 UN: 3261 ADR: 8,III |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | min. 99.0 % |
Melting point | 83.0 to 88.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...