Stampa…
Chromone-3-carboxaldehyde ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms: 4-Oxo-4H-chromene-3-carbaldehyde
O0291-25G O0291-5G
TCIAO0291-25GEA 91 EUR
TCIAO0291-25G TCIAO0291-5G
Chromone-3-carboxaldehyde ≥98.0% (by GC)
Chromone-3-carboxaldehyde
Formula: C₁₀H₆O₃
Peso molecolare: 174,16 g/mol
MDL Number: MFCD00014667
CAS Number: 17422-74-1
EINECS: 241-451-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2eda207c51e034f27a94e234b91b0e6159e1610a&vid=2eda207c51e034f27a94e234b91b0e6159e1610a&prid=web&clid=SAPDAM
Visualizza i codici
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 154.0 deg-C