Stampa…

1-Nitro-9,10-anthraquinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 1-Nitroanthraquinone, 1-Nitroanthracene-9,10-dione

N0127-25G N0127-500G
TCIAN0127-25GEA 37 EUR
TCIAN0127-25G TCIAN0127-500G
1-Nitro-9,10-anthraquinone ≥98.0% (by GC)
1-Nitro-9,10-anthraquinone
Formula: C₁₄H₇NO₄
Peso molecolare: 253,2097 g/mol
MDL Number: MFCD00019140
CAS Number: 82-34-8
EINECS: 201-413-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1c4add5681d5b179854e8659b79bee36104ce142&vid=1c4add5681d5b179854e8659b79bee36104ce142&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 230.0 to 234.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR