trans-4-Nitrostilbene ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₄H₁₁NO₂ |
MDL Number:
MFCD00053614 CAS Number: 1694-20-8 EINECS: 223-653-5 |
|
Risultati del test di specificazione
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 155.0 to 159.0 deg-C |
Solubility in hot Toluene | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...