4-Bromo-trans-stilbene ≥98.0% (by GC)
Fornitore: TCI EUROPE
Synonyms:
trans-4-Bromostilbene
B2416.0005
B2416-1G
B2416-5G
TCIAB2416.0005EA
0
EUR
TCIAB2416.0005
TCIAB2416-1G
TCIAB2416-5G
4-Bromo-trans-stilbene ≥98.0% (by GC)
4-Bromo-trans-stilbene
Formula:
C₁₄H₁₁Br Peso molecolare: 259,15 g/mol |
MDL Number:
MFCD00155020 CAS Number: 13041-70-8 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 138.0 to 142.0 deg-C |
Solubility in Toluene | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...