2-(1-Adamantyl)-4-bromoanisole ≥98.0% (by GC)
Fornitore: TCI
Formula:
C₁₇H₂₁BrO |
MDL Number:
MFCD03855308 CAS Number: 104224-63-7 |
|
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...