Stampa…

2-Amino-4-(1-naphthyl)thiazole 97%

Fornitore: Thermo Fisher Scientific
Attenzione

B23882.03 B23882.06
B23882.03EA 47.6 EUR
B23882.03 B23882.06
2-Amino-4-(1-naphthyl)thiazole 97%
2-Amino-4-(1-naphthyl)thiazole

Purity: 97% CAS No.: 56503-96-9 Molecular Formula: C13H10N2S Formula Weight: 226.30 Melting Point: 155-158° Beilstein Reference: 164858

Formula: C₁₃H₁₀N₂S
Punto di fusione: 155…158 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00236016
CAS Number: 56503-96-9

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR