O-Methylisourea hemisulphate ≥99%
Fornitore: Thermo Scientific
Synonyms:
O-Methylisouronium sulphate
B22036.0B
B22036.18
B22036.30
B22036.0BEA
536
EUR
B22036.0B
B22036.18
B22036.30
O-Methylisourea hemisulphate ≥99%
O-Methylisourea hemisulphate
Purity: 99%
CAS No.: 52328-05-9
Molecular Formula: C2H6N2O.0.5H2SO4
Formula Weight: 123.12
Melting Point: 163-167°
MDL No.: MFCD00040594
Formula:
C₄H₁₄N₄O₆S Peso molecolare: 246,24 g/mol Punto di fusione: 163…167 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00040594 CAS Number: 52328-05-9 EINECS: 257-851-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...