Stampa…

Isosorbide ≥98.0% (by GC)

Fornitore: TCI

Synonyms: D-Isosorbide, (3R,3aR,6S,6aR)-Hexahydrofuro[3,2-b]furan-3,6-diol

I0407-25G I0407-500G I0407-100G
TCIAI0407-25GEA 16 EUR
TCIAI0407-25G TCIAI0407-500G TCIAI0407-100G
Isosorbide ≥98.0% (by GC)
Isosorbide
Formula: C₆H₁₀O₄
Peso molecolare: 146,14 g/mol
Punto di ebollizione: 175°C
MDL Number: MFCD00064827
CAS Number: 652-67-5
EINECS: 211-492-3
Merck Index: 14,05224
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f54ffe893107ed94d94f9512127ee666a3af10e0&vid=f54ffe893107ed94d94f9512127ee666a3af10e0&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 58.0 to 63.0 deg-C
Specific rotation [a]20/D +43.0 to +47.0 deg (C=5, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR