Ethyl-3-methyl-3-phenylglycidate (mixture of cis and trans isomers) ≥95.0% (by GC)
Fornitore: TCI
|
Attenzione
|
Synonyms:
Ethyl 3-Methyl-3-phenylglycidate (mixture of isomers), Ethylmethylphenylglycidate Ethyl 3-methyl-3-phenylglycidate 3-Methyl-3-phenyl glycidic acid ethyl ester, Ethyl 2,3-Epoxy-3-phenylbutyrate (mixture of isomers), 2,3-Epoxy-3-phenylbutyric acid Ethyl Ester (mixture of isomers), 3-Methyl-3-phenylglycidic acid Ethyl Ester (mixture of isomers)
Formula:
C₁₂H₁₄O₃ Punto di ebollizione: 86°C Flash Pt: 134°C |
MDL Number:
MFCD00022338 CAS Number: 77-83-8 EINECS: 201-061-8 |
|
Risultati del test di specificazione
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % (total of isomer) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...