Stampa…
HPLC columns, NUCLEOSIL® C18
Fornitore: MACHEREY-NAGEL
720041.40 720051.40 720730.46 720740.46 720949.46 721070.30 721074.20 721074.30 721075.30 720002.20 720002.30 720002.40 720002.46 720014.20 720014.30 720014.40 720014.46 720018.40 720018.46 720023.40 720023.46 720040.40 720040.46 720041.46 720042.40 720043.40 720043.46 720051.46 720055.40 720055.46 720065.40 720065.46 720074.46 720077.46 720098.46 720120.46 720133.40 720133.46 720150.40 720150.46 720120.40 720841.46 720149.46 720141.46 720141.40 720110.46 720951.46 720149.40 721022.30 720140.46 720701.46 721085.30 721473.30
MANA720041.40EA 450 EUR
MANA720041.40 MANA720051.40 MANA720730.46 MANA720740.46 MANA720949.46 MANA721070.30 554-5795 554-5796 MANA721075.30 MANA720002.20 MANA720002.30 MANA720002.40 MANA720002.46 554-5791 554-5792 554-5793 554-5794 MANA720018.40 MANA720018.46 MANA720023.40 MANA720023.46 MANA720040.40 MANA720040.46 MANA720041.46 MANA720042.40 MANA720043.40 MANA720043.46 MANA720051.46 MANA720055.40 MANA720055.46 MANA720065.40 MANA720065.46 MANA720074.46 MANA720077.46 MANA720098.46 554-5790 MANA720133.40 MANA720133.46 MANA720150.40 MANA720150.46 554-5789 MANA720841.46 MANA720149.46 MANA720141.46 MANA720141.40 MANA720110.46 MANA720951.46 MANA720149.40 MANA721022.30 MANA720140.46 MANA720701.46 MANA721085.30 MANA721473.30
HPLC columns, NUCLEOSIL® C18
Colonne cromatografiche
NUCLEOSIL® C18 are octadecyl modified silica phases for HPLC.
  • Robust spherical silica gel
  • Hydrophobic, weakly polar, endcapped phases for RP chromatography
  • Separation mechanism based on hydrophobic (van der Waals) and slight residual silanol interactions
EC guard columns are used for analytical EC columns with the column protection system guard column holder (554-5851).
Confezione: Standard NUCLEOSIL® C18 phases are available as packed EC columns for analytical HPLC.
Visualizza i codici
Specifiche
Environmentally Preferable
Carico di carbonio 6,5% (300 Å) 2% (500 Å) 1% (1000 Å) 14,5% (50 Å) 15% (100 Å) 11% (120 Å)
Con endcapping
Range di pH 2 - 8 2 - 8 2 - 8 2 - 8 2 - 8 2 - 8
Codice USP L1 L1 L1 L1 L1 L1